missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with PTFE Membrane and Minispike Outlet

Universal membrane syringe filters for all of your analytical filtration requirements.

Brand:  Cytiva (formerly Pall Lab) 4555

Additional Details : Weight : 0.72200kg

 View more versions of this product

Product Code. 17104361

  • 2275.00 EUR / Pack of 1000
Estimated Shipment: 03-07-2024
to see stock.
Add to basket

Description

Description

  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

PTFE
Non-sterile
<14 μL
1000/Pk.
100°C
0.45 μm
1 cm2
Polypropylene
Minispike Outlet
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers