Learn More
Honeywell Fluka™ Acetate Buffer Solution Ph 4.6, Honeywell Fluka™
Sodium acetate / acetic acid, for complexometry
Brand:  Honeywell Fluka™ 31048-500ML
44.16 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : CAS Number : 126-96-5 Weight : 0.50500kg
 View more versions of this product
Product Code. 15633190
  • 46.48 EUR / 500mL
Estimated Shipment: 10-06-2024
Description
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
126-96-5
500mL