Learn More
Cytiva Acetal plastic/Fluroro Rubber/Nitrile Rubber/Polypropylene/Superpolyxymethylene RC 26 Packing Reservoir
Product Code. 10871081 Shop All Cytiva Products
Click to view available options
Packaging Quantity:
1 Each
Unit Size:
Each
Product Code. 10871081
Brand: Cytiva 19520801
Only null left
Add to basket
Add to basket
Quick and easy to assemble
RC packing reservoirs, which are available for all three C column types, are quick and easy to assemble. Curved walls minimise the separation of matrix beads by size during packing to produce homogeneous columns. Once the matrix is in the column, the RC reservoir can be connected to a pump or Mariotte flask to give optimal flow rates for column packing. Packing reservoirs for each column diameter optimise packing conditions.
TRUSTED_SUSTAINABILITY
Specifications
Type RC 26 Packing Reservoir
Packaging Quantity 1 Each
Material Acetal, Fluoro Rubber, Nitrile Rubber, Polypropylene, Superpolyxymethylene
Pressure 14.5 psi