Learn More
Cytiva Acetal plastic/Fluroro Rubber/Nitrile Rubber/Polypropylene/Superpolyxymethylene RC 26 Packing Reservoir
Shop All Cytiva Products
Click to view available options
Packaging Quantity:
1 Each
Unit Size:
Each
Description
RC packing reservoirs, which are available for all three C column types, are quick and easy to assemble. Curved walls minimise the separation of matrix beads by size during packing to produce homogeneous columns. Once the matrix is in the column, the RC reservoir can be connected to a pump or Mariotte flask to give optimal flow rates for column packing. Packing reservoirs for each column diameter optimise packing conditions.
Specifications
Specifications
| Type | RC 26 Packing Reservoir |
| Packaging Quantity | 1 Each |
| Material | Acetal, Fluoro Rubber, Nitrile Rubber, Polypropylene, Superpolyxymethylene |
| Pressure | 14.5 psi |