Learn More
Digi-Sense™ Accessories for the 20250-40 Digi-Sense™ Temperature Humidity Touch-screen Recorder
Product Code. 15820016
Click to view available options
Product Type:
Extension Sensor Cable
Replacement Power Supply
Replacement Temperature/RH Sensor
Replacement USB Cable
Unit Size:
Each
Product Code. 15820016
Brand: Digi-Sense™ 2025047
Only null left
Add to basket
Add to basket
The touch-screen reader has 8GB on board memory to provide virtually unlimited data logging capacity
  • Includes A2LA accredited NIST-Traceable Calibration Certificate
  • Measures, logs, and displays temperature, humidity, and dew point
  • Removable sensor for local or remote measurements using extension cable included
  • Wall mount or desk mount operation
  • Resident self-booting software, no installation necessary
  • Comes with NIST calibration
  • Stored data downloads to computer via USB cable provided in CSV format
Warranty
2 Years
Specifications
For Use With (Application) Extension Cable for Temperature/RH Touch Screen Recorder with NIST Traceable Calibration
Product Type Extension Sensor Cable
For Use With (Equipment) Temperature/RH Touch Screen Recorder with NIST Traceable Calibration (20250-40)