Learn More
Labconco™ Accessories for Labconco™ Fiber Glass Desiccator
Vacuum Tubing Kit allows connection of Vacuum Desiccator's valve to an accessory pump.
272.00 EUR
Specifications
For Use With (Equipment) Labconco Fiber Glass Desiccator
View More Specs
Products 1
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
11791379
View Documents Promotion Details
Labconco™
5530600
Stacking Rack
272.00 EUR
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-06-2024
Description
Vacuum Tubing Kit includes 1/4 in. ID and 1/2 in. ID vacuum tubing, each 48 in. long, two each 1/4 in. and 1/2 in. tubing clamps; 1/4 in. to 1/2 in. barbed reducer. Kit allows connection of Labconco Vacuum Desiccator's 1/4 in. diameter brass needle valve to an accessory pump with 1/2 in. connector.
Order Info
Dimensions: 12L x 12 in. W
Specifications
Specifications
Labconco Fiber Glass Desiccator