Learn More
Fisherbrand™ Accessories for Fisherbrand™ RT Basic Series Magnetic Stirrers
Optimize performance and flexibility
Brand:  Fisherbrand™
9.80 EUR valid until 2018-12-31
Use promo code "13295" to get your promotional price.
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15396607
  • 12.65 EUR / Each
Estimated Shipment
16-01-2019
Add to basket
Description
Description
  • White
  • Silicone
Specifications
Specifications
120mm
RT Basic Series Magnetic Stirrer
White silicone plate cover
Videos
Documentation Special Offers
Special Offers
  • Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    : 13295
    2018-12-31
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/09/2018 and 31/12/2018. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions