missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TNNI3 (Rabbit) ELISA Kit
Enzyme Immunoassay for the quantitative measurement of TNNI3 in serum
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | HRP |
Product Type | TNNI3 (Rabbit) ELISA Kit |
Quantity | 96 Assays |
Regulatory Status | For research use only (RUO) |
Specifications
Colorimetric | |
TNNI3 (Rabbit) ELISA Kit | |
For research use only (RUO) |
HRP | |
96 Assays |