missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TNNI3 (Rabbit) ELISA Kit

Enzyme Immunoassay for the quantitative measurement of TNNI3 in serum

Brand:  Abnova™ KA2487.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007325

  • 732.00 EUR / 1 set


Estimated Shipment
12-02-2020
Add to basket

Specifications

Specifications

HRP
Colorimetric
Anti cTnI-coated microtiter plate (96 wells), Rabbit cTnI (Stock Lyophilized), cTnI Diluent (25mL), cTnI HRP Conjugate (11mL), 20x Wash Solution (50mL), TMB Reagent (11mL), Stop Solution (1N HCl) (11mL)
96 Assays
Store Lyophilized Standard at -20°C. Store all other components at 4°C.
200μL
TNNI3 (Rabbit) ELISA Kit is for the quantitative measurement of TNNI3 in plasma and serum.
Quantification
TNNI3 (Rabbit) ELISA Kit
For research use only (RUO)
96 Assays
Videos
Documents