missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TNF (Rat) ELISA Kit

Brand:  Abnova™ KA3115.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007855

  • 578.00 EUR / 1 set


Estimated Shipment
07-08-2020
Add to basket

Includes: One 96-well plate pre-coated with anti-Rat TNFα antibody, 96 (8 x 12) wells
Lyophilized TNFα standards, 10ng x 2
Sample/Standard diluent buffer, 30mL
Biotin conjugated anti-Rat TNFα antibody (Concentrated), Dilution: 1:100, 130μL
Antibody diluent buffer, 12mL
Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
ABC diluent buffer, 12mL
TMB substrate, 10mL
Stop solution, 10mL
Wash buffer (25X), 30mL

Description

Description

This kit is based on sandwich enzyme-linked immune-sorbent assay technology
Specifications

Specifications

15.6 to 1000pg/mL (Body Fluid, Tissue Lysate, Cell Culture Supernatant), 7.8 to 501pg/mL (Serum)
Rat
1pg/mL
Quantification
One 96-well plate pre-coated with anti-Rat TNFα antibody 96 (8x12) wells, Lyophilized anti-Rat TNFα standards (10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Rat TNFα antibody (Concentrated), Dilution: 1:100 (130μL) Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
For research use only (RUO)
100μL
Store at 4°C for 4 months, or at -20°C for 8 months.
100μL
Biotin
Tnf (Rat) ELISA Kit is intended for the quantitative measurement of rat Tnf.
Colorimetric
96 wells
Tnf (Rat) ELISA Kit
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
<1pg/mL
96 Assays
Videos
Documents