missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SERPINA1 (Human) ELISA Kit

Brand:  Abnova™ KA0458.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006105

  • 578.00 EUR / 1 set


Estimated Shipment
17-03-2020
Add to basket

Includes: Human alpha-1 Antitrypsin Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against human A1AT
Sealing Tapes
Human alpha-1 Antitrypsin Standard: Human A1AT in a buffered protein base, lyophilized, 120µg
Biotinylated Human alpha-1 Antitrypsin: lyophilized
MIX Diluent Concentrate (10x)
Wash Buffer Concentrate (20x)
Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
Stop Solution: A 0.5N hydrochloric acid

Description

Description

This assay employs a quantitative competitive enzyme immunoassay technique that measures human A1AT in less than 3 hours
Specifications

Specifications

0.039 to 40μg/mL
Human
0.018μg/mL
Quantification
Antibody coated microwells 96 wells, Human A1AT Standard 20μg, Biotinylated A1AT (1 vial), MIX Diluent Concentrate (10x) (30mL), Wash Buffer Concentrate (20x) (30mL), Streptavidin-Peroxidase Conjugate (SP Conjugate) 90Chromogen Substrate (8mlL), Stop Solution: (12mL)
ELISA Kit
For research use only (RUO)
25μL
96 Assays
Biotin
SERPINA1 (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human SERPINA1 in plasma and serum.
Colorimetric
96 wells
SERPINA1 (Human) ELISA Kit
1 Kit
Plasma, Serum
Store components of the kit at 4°C or -20°C as described in the protocol.
25μL
Videos
Documents