missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Serotonin ELISA Kit

Enzyme immunoassay for the quantitative determination of Serotonin in serum, urine and platelets

Brand:  Abnova™ KA2518.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007355

  • 578.00 EUR / 1 set


Estimated Shipment
12-02-2020
Add to basket

Description

Description

  • Sample Type: Platelet, Serum, Urine
  • Sample Volume: 25μL

Applications:

Quantification

Specifications

Specifications

Serotonin ELISA Kit is an ultra-sensitive Enzyme Immunoassay for the quantitative determination of Serotonin.
Quantification
Serotonin ELISA kit
Biological sample
Store at 4°C.
Colorimetric
Serotonin ELISA Kit
1 Kit
100μL
Videos
Documents