missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Sell (Rat) ELISA Kit

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Rat Sell

Brand:  Abnova™ KA1830.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006865

  • 578.00 EUR / 1 set


Estimated Shipment
19-02-2020
Add to basket

Specifications

Specifications

Biotin
Sell (Rat) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of rat Sell.
Colorimetric
96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB one step Substrate reagent, 8mL stop solut
96 Assays
Cell Culture Supernatant, Plasma, Serum
96 Assays
Rat
6pg/mL
Quantification
Sell (Rat) ELISA Kit
RUO
Store the kit at -20°C. Standard should be stored at -20°C or -80°C. Avoid repeated freezing and thawing.
100μL
Videos
Documents