missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Rubella virus IgG ELISA Kit

solid phase Enzyme Immunoassay for the quantitative measurement of IgG class autoantibodies against Rubella virus.

Brand:  Abnova™ KA0223.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16005955

  • 263.00 EUR / 1 set


Estimated Shipment
16-10-2020
Add to basket

Specifications

Specifications

HRP
Rubella IgG ELISA Kit is an enzyme immunoassay for the quantitative measurement of Rubella IgG.
Quantification
Rubella virus IgG ELISA Kit
1 Kit
Serum
96 Assays
Viruses
Colorimetric
96 wells-Microtiter wells Rubella antigen-coated wells, Enzyme Conjugate Reagent (12mL), Calibrator Set (150uL), Positive Control (100uL), Negative Control (100uL), Samples Diluent (22mL), Cut-off Calibrator (100uL), 20X Wash Buffer (50mL),Positive calibrator (100uL), Negative calibrator (100uL), TMB Reagent (11mL), Stop Solution (11mL)
Rubella Virus IgG ELISA kit
RUO
Store the kit at 4°C.
100μL
Videos
Documents