missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Retn (Mouse) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Retn

Brand:  Abnova™ KA0995.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006475

  • 578.00 EUR / 1 set


Estimated Shipment
20-02-2020
Add to basket

Specifications

Specifications

156 to 10000pg/mL
Mouse
3pg/mL
Quantification
Retn (Mouse) ELISA Kit
1 Kit
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
96 Assays
Biotin
Retn (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Retn.
Colorimetric
96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Mouse Retn standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Mouse Retn Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin -Biotin
Retn (Mouse) ELISA kit
RUO
Store the kit at-20°C.
100μL
Videos
Documents