missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Retn (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Retn
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | Biotin |
Cross Reactivity | Mouse |
Product Type | Retn (Mouse) ELISA kit |
Quantity | 1 Kit |
Specifications
Colorimetric | |
Mouse | |
1 Kit | |
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate |
Biotin | |
Retn (Mouse) ELISA kit | |
For research use only (RUO) | |
100 μL |