missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Retn (Mouse) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Retn

Brand:  Abnova™ KA3104.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007845

  • 578.00 EUR / 1 set


Estimated Shipment
31-08-2020
Add to basket

Specifications

Specifications

31.2 to 2000pg/mL
Mouse
2pg/mL
Quantification
Retn (Mouse) ELISA Kit
RUO
Store at 4°C for 4 months, or at -20°C for 8 months.
100μL
Biotin
Retn (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Retn.
Colorimetric
96-well plate pre-coated with anti-Mouse Resistin antibody, Lyophilized Resistin standards (100ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Mouse Resistin antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated) (130uL), ABC diluent buffer (12mL),TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X)(30mL)
96 Assays
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
96 Assays
Videos
Documents