missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PTX3 (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human PTX3

Brand:  Abnova™ KA1007.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006485

  • 1858.00 EUR / 1 set


Estimated Shipment
11-02-2020
Add to basket

Specifications

Specifications

0.5 to 20ng/mL
Human
0.1ng/mL
Quantification
PTX3 (Human) ELISA Kit
1 Kit
Cell Culture Supernatant, Plasma
96 Assays
HRP
PTX3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of PTX3.
Colorimetric
96-well Antibody coated microwell, 12mL HRP Conjugated Anti-PTX3 Antibody reagent, 0.1mL x 7 vials Human PTX3 standard (0, 0.5, 1.0, 2.5, 5.0, 10, 20ng/mL), 100mL Wash Buffer 10X concentrate, 15mL Dilution Buffer, 12mL TMB solution, 12mL stop solution
PTX3 (Human) ELISA kit
RUO
Store the kit at 4°C.
10μL
Videos
Documents