missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PTX3 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human PTX3
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Human |
Product Type | PTX3 (Human) ELISA kit |
Quantity | 1 Kit |
Regulatory Status | For research use only (RUO) |
Specifications
Colorimetric | |
PTX3 (Human) ELISA kit | |
For research use only (RUO) |
Human | |
1 Kit |