missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Prostaglandin F2 alpha ELISA Kit

Brand:  Abnova™ KA0310.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006025

  • 475.00 EUR / 1 set


Estimated Shipment
06-08-2020
Add to basket

Includes: Donkey anti-Sheep IgG Microtiter Plate, 96 Wells
PGF2α ELISA Conjugate, 5mL
PGF2α EIA Antibody, 5mL
Assay Buffer, 30mL
Wash Buffer Concentrate, 27mL
Prostaglandin F2α Standard, 0.5mL
pNpp Substrate, 20mL
Stop Solution, 5mL
Plate Sealer, 1 each

Description

Description

The Prostaglandin F2α kit is a competitive immunoassay for the quantitative determination of Prostaglandin F2α in biological fluids
Specifications

Specifications

3.05 to 50000pg/mL
PGF2-alpha ELISA Kit is a competitive immunoassay for the quantitative determination of Prostaglandin F2-alpha in biological fluids.
Colorimetric
96 wells
Prostaglandin F2 alpha ELISA Kit
1 Kit
Cell Culture Supernatant, Plasma, Saliva, Serum, Urine
6.71pg/mL
96 Assays
AP
6.71pg/mL
Quantification
Microwells (96 wells), EIA Conjugate (5mL), EIA Antibody (5mL), Assay Buffer (30mL), Wash Buffer Concentrate (30mL), Standard (0.5mL), pNpp Substrate (20mL), Stop Solution: (5mL)
ELISA Kit
For research use only (RUO)
100μL
Store the kit at 4°C.
100μL
Videos
Documents