missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Prostaglandin F2 alpha ELISA Kit
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | AP |
Product Type | ELISA Kit |
Includes | Microwells (96 wells), EIA Conjugate (5mL), EIA Antibody (5mL), Assay Buffer (30mL), Wash Buffer Concentrate (30mL), Standard (0.5mL), pNpp Substrate (20mL), Stop Solution: (5mL) |
Regulatory Status | For research use only (RUO) |
Description
The Prostaglandin F2 alpha ELISA Kit is a competitive immunoassay for the quantitative determination of Prostaglandin F2α.Specifications
Colorimetric | |
ELISA Kit | |
For research use only (RUO) |
AP | |
Microwells (96 wells), EIA Conjugate (5mL), EIA Antibody (5mL), Assay Buffer (30mL), Wash Buffer Concentrate (30mL), Standard (0.5mL), pNpp Substrate (20mL), Stop Solution: (5mL) | |
Store the kit at 4°C |