Learn More
Abnova™ Progesterone ELISA Kit
Brand:  Abnova™ KA3392.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16007975
  • 366.00 EUR / 1 set
Estimated Shipment
11-02-2020
Add to basket
Specifications
Specifications
0 to 3200ng/mL
52.9pg/mL
Quantitative detection of Progesterone in extracted dried fecal samples, urine and tissue culture media samples
Progesterone ELISA Kit
Dried Fecal Extract, Tissue Culture Medium, Urine
4°C
50μL
HRP
Colorimetric
Coated Clear 96 Well Plates: Clear plastic microtiter plate coated with goat anti-mouse IgG (1 plate), Progesterone Standard: Progesterone at 32,000pg/mL in a special stabilizing solution (125mL), Progesterone Antibody: A mouse monoclonal antibody specific for progesterone (3mL), Progesterone Conjugate: A progesterone-peroxidase conjugate in a special stabilizing solution (3mL), Assay Buffer Concentrate: A 5X concentrate that should be diluted with deionized or distilled water (28mL), Wash Buffer Concentrate: A 20X concentrate that should be diluted with deionized or distilled water (30mL), TMB Substrate (11mL), Stop Solution: A 1 M solution of hydrochloric acid. CAUSTIC (5mL), Plate Sealer (1 slice)
RUO
50μL
96 Assays