missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PGF (Human) ELISA Kit
In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human PGF
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Human |
Product Type | PGF (Human) ELISA Kit |
Quantity | 96 Assays |
Regulatory Status | For research use only (RUO) |
Specifications
Colorimetric | |
PGF (Human) ELISA Kit | |
For research use only (RUO) |
Human | |
96 Assays |