missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PGF (Human) ELISA Kit

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human PGF

Brand:  Abnova™ KA0410.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006075

  • 578.00 EUR / 1 set


Estimated Shipment
31-08-2020
Add to basket

Specifications

Specifications

15.6 to 1000pg/mL
Human
1pg/mL
Quantification
PGF (Human) ELISA Kit
1 Kit
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
96 Assays
Biotin
PGF (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human PGF.
Colorimetric
96-well plate pre-coated with anti-Human PGF antibody, Lyophilized PGF standards (10ng x 2), Sample diluent buffer (30mL), Biotin conjugated anti-Human PGF antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (130uL), Antibody diluent buffer (12mL), ABC diluent buffer (12mL), TMB color developing agent (10mL), Stop solution (10mL)
PGF (Human) ELISA kit
RUO
Store the kit at-20°C.
100μL
Videos
Documents