Learn More
Abnova™ ORM1 (Monkey) ELISA Kit
Brand:  Abnova™ KA2439.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16007285
  • 578.00 EUR / 1 set
Estimated Shipment
06-08-2020
Add to basket
Description
Description
ORM1 (Monkey) ELISA Kit is an enzyme immunoassay for the quantitative determination of ORM1 in monkey serum or plasma.
Quantification
Specifications
Specifications
ORM1 (Monkey) ELISA Kit is for the quantitative measurement of ORM1 in serum.
Quantification
ORM1 (Monkey) ELISA Kit
Store the kit at 4°C.