missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ LGALS3 (Mouse) ELISA Kit

Brand:  Abnova™ KA2147.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007075

  • 578.00 EUR / 1 set


Estimated Shipment
28-08-2020
Add to basket

Includes: Galectin-3 Microplate (Item A): 96 wells coated with anti-mouse Galectin-3
Wash Buffer Concentrate (20x) (Item B)
Standards (Item C): recombinant mouse Galectin-3
Assay Diluent A (Item D)
Assay Diluent B (Item E)
Detection Antibody Galectin-3 (Item F): 2 vial of biotinylated antimouse Galectin-3 (each vial is enough to assay half microplate
HRP-Streptavidin Concentrate (Item G): 8µL 20,000x concentrated HRP-conjugated streptavidin
TMB One-Step Substrate Reagent (Item H): 12mL of 3,3′,5,5′-tetramethylbenzidine (TMB) in buffer solution
Stop Solution (Item I): 2M sulfuric acid

Description

Description

This assay employs an antibody specific for mouse Galectin-3 coated on a 96-well plate
Specifications

Specifications

HRP
Lgals3 (Mouse) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of mouse Lgals3.
Colorimetric
96 wells
Lgals3 (Mouse) ELISA Kit
Serum, Plasma, Cell Culture Supernatant, Urine
6pg/mL
96 Assays
Mouse
6pg/mL
Quantification
Antibody coated microwells (96 wells), Wash Buffer Cincentrate (20x) (ItemB): 25mL, Standard (Item C): 2 vials, Assay Diluent A (Item D): 30mL, Assay Diluent B (Item E): 15mL, Detection Antibody MSP (Item F): 2 vials, HRP-Streptavidian concentrrate (ItemG): 8μL, TMB One-Step Substrate Reagent (ItemH): 13mL, Stop solution (Item I): 8mL
RUO
100μL
Store the kit at -20°C.
100μL
Videos
Documents