missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LGALS3 (Mouse) ELISA Kit
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Mouse |
Product Type | Lgals3 (Mouse) ELISA Kit |
Regulatory Status | For research use only (RUO) |
Content And Storage | Store the kit at -20°C |
Description
Lgals3 (Mouse) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technologySpecifications
Colorimetric | |
Lgals3 (Mouse) ELISA Kit | |
Store the kit at -20°C |
Mouse | |
For research use only (RUO) | |
100 μL |