missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IL6 (Mouse) ELISA Kit

Brand:  Abnova™ KA2354.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007265

  • 578.00 EUR / 1 set


Estimated Shipment
11-02-2020
Add to basket

Includes: 96-well plate precoated with anti-mouse IL-6 antibody, 96 (8x12) wells
Lyophilized recombinant mouse IL-6 standard, 10ng/tube x 2
Biotinylated anti-mouse IL-6 antibody (dilution 1:100), 130μL
Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
Sample diluent buffer, 30mL
Antibody diluent buffer, 12mL
ABC diluent buffer, 12mL
TMB color developing agent, 10mL
TMB stop solution, 10mL

Description

Description

Il6 (Mouse) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology
Specifications

Specifications

15.6 to 1000pg/mL
Mouse
<1pg/mL
Quantification
Lyophilized recombinant mouse Il6 standard (10ng/tube x 2), One 96-well plate precoated with anti-mouse Il6 antibody (1), Sample diluent buffer (30mL), Biotinylatedanti-mouse Il6 antibody, dilution 1:100 (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100 (130μL), ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
ELISA Kit
RUO
100μL
Natural and recombinant mouse IL-6
96 Assays
Biotin
Il6 (Mouse) ELISA Kit is an immunoassay for quantitative detection of mouse IL 6 in serum, plasma, body fluid, tissue lysate or cell culture supernate.
Colorimetric
96 wells
Il6 (Mouse) ELISA Kit
1 Kit
Cell culture supernatant, Plasma (Citrate, EDTA, heparin), Serum
<1pg/mL
Store the kit at-20°C.
100μL
Videos
Documents