missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IL20 (Human) ELISA Kit

Brand:  Abnova™ KA2191.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007115

  • 578.00 EUR / 1 set


Estimated Shipment
31-07-2020
Add to basket

Includes: Microplate or Strips Coated w/Capture Antibody, 12 x 8 Strips
Ready-to-Use Streptavidin-HRP, 12mL, Clear container
Ready-to-Use Substrate, 12mL, Brown container
Stop Solution, 12mL, Clear container
Wash Buffer (10x), 50mL, Clear container
Protein Standard Diluent. 12mL, Clear container
Sample Diluent, 12mL, Clear container
Detection Antibody Diluent, 12mL, Clear container
Biotin-Labeled Detection Antibody, Lyophilized, Yellow container
Protein Standard, Lyophilized (100ng), Red container
Adhesive Plate Sealers, 4 Sheets

Description

Description

This immunoassay utilizes the quantitative technique of a “Sandwich” Enzyme-Linked Immunosorbent Assay (ELISA) where the target protein (antigen) is bound in a “sandwich” format by the primary capture antibodies coated to each well-bottom and the secondary detection antibodies added subsequently by the investigator
Specifications

Specifications

Not Applicable
Human
Colorimetric
96 wells
IL20 (Human) ELISA Kit
1 Kit
Cell Lysate, Plasma, Serum
16-4000pg/mL
96 assays
Biotin
IL20 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL20.
Quantification
Antibody coated microwells (96 wells), Biotin-conjugated detection antibody (1 vial), Avidin-HRP conjugate (10μL), Cytokine protein standard (1 vial); Ready-to-use substrate (11mL), Stop solution (11mL), Wash buffer 10X (50mL), Protein standard diluent (15mL), Detection antibody diluent (15mL), HRP diluent (15mL)
ELISA Kit
RUO
100μL
Store the kit at 4°C.
100μL
Videos
Documents