missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL20 (Human) ELISA Kit
Use to measure quantitatively human IL20. Sandwich enzyme immunoassay.
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | Biotin |
Cross Reactivity | Human |
Product Type | ELISA Kit |
Regulatory Status | For research use only (RUO) |
Description
IL20 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL20.Specifications
Colorimetric | |
Human | |
For research use only (RUO) |
Biotin | |
ELISA Kit | |
100 μL |