missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Il10 (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse IL-10
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Mouse |
Product Type | Il10 (Mouse) ELISA Kit |
For Use With (Application) | Quantification |
Quantity | 1 Kit |
Specifications
Colorimetric | |
Il10 (Mouse) ELISA Kit | |
1 Kit |
Mouse | |
Quantification | |
For research use only (RUO) |