missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Il10 (Mouse) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse IL-10

Brand:  Abnova™ KA3070.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007815

  • 578.00 EUR / 1 set


Estimated Shipment
20-02-2020
Add to basket

Specifications

Specifications

15.6 to 1000pg/mL
Mouse
1pg/mL
Quantification
Il10 (Mouse) ELISA Kit
RUO
Store at 4°C for 4 months, or at -20°C for 8 months.
100μL
Biotin
Il10 (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Il10.
Colorimetric
96-well plate pre-coated with anti-Mouse IL-10 antibody, Lyophilized IL-10 standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Mouse IL-10 antibody (Concentrated Dilution: 1:100) (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated Dilution: 1:100) (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
1 Kit
Body Fluid, Cell Culture Supernate, Serum, Tissue Lysate
96 Assays
Videos
Documents