missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IL10 (Human), Biotin, ELISA Kit

IL10 (Human) ELISA Kit is intended for the quantitative measurement of human IL10.

Brand:  Abnova™ KA0125.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16005875

  • 1860.00 EUR / 1 set


Estimated Shipment
20-02-2020
Add to basket

Includes:
  • One 96-well plate pre-coated with anti-Human IL-10 antibody 96 (8x12) wells
  • Lyophilized IL-10 standards 10ng x 2
  • Sample / Standard diluent buffer 30mL
  • Biotin conjugated anti-Human IL-10 antibody (Concentrated), Dilution: 1:100 130 ul
  • antibody diluent buffer 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 130 ul
  • ABC diluent buffer 12mL
  • TMB substrate 10mL
  • Stop solution 10mL
  • Wash buffer (25X) 30mL

Description

Description

  • This kit was based on sandwich enzyme-linked immune-sorbent assay technology
  • Anti-IL-10 polyclonal antibody was pre-coated onto 96-well plates
  • And the biotin conjugated anti-IL-10 polyclonal antibody was used as detection antibodies
  • Store at 2-8°C for 4 months, or at -20°C for 8 months
  • For research use only

Specifications

Specifications

3.1 to 200pg/mL
IL10 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL-10.
IL10 (Human) ELISA Kit
Store the controls at -20°C. Others at 4°C. Avoid repeated freezing and thawing.
Human
Biotin
Quantification
RUO
1 Kit
Serum
Videos
Documents