missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IgA (Rat) ELISA Kit

Enzyme-linked immunoassay for the quantitative determination of IgA in Rat biological samples

Brand:  Abnova™ KA1950.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006965

  • 366.00 EUR / 1 set


Estimated Shipment
19-03-2020
Add to basket

Specifications

Specifications

31.2 to 1000ng/mL
Rat
Colorimetric
50mL Diluent concentrate, 50mL wash solution concentrate, 150μL Enzyme Antibody Conjugate concentrate, 12mL TMB Substrate solution, 12mL stop solution, 96-well microtiter plate, 1 vial calibrator (lyophilized)
IgA (Rat) ELISA kit
For research use only (RUO)
Store the kit at 4°C.
100μL
HRP
IgA (Rat) ELISA Kit is a highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of IgA in rat biological samples.
Quantification
IgA (Rat) ELISA Kit
1 Kit
Plasma, Serum
96 Assays
Videos
Documents