missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ICAM1 (Human) ELISA Kit

Brand:  Abnova™ KA0113.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16005865

  • 578.00 EUR / 1 set


Estimated Shipment
11-02-2020
Add to basket

Includes: Microwell Plate coated with monoclonal antibody to human sICAM-1, 96 (8 x 12) wells
HRP-Conjugate anti-human sICAM-1 monoclonal antibody, 100µL
human sICAM-1 Standard, 100ng/mL, 500µL x2
Control high, lyophilized, 1 vial
Control low, lyophilized, 1 vial
Sample diluent, 12mL
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), 5mL
Wash Buffer Concentrate 20x (PBS with 1% Tween 20), 50mL
Substrate Solution (tetramethyl-benzidine), 15mL
Stop Solution (1M Phosphoric acid), 15mL
Blue-Dye, 0.4mL
Green-Dye, 0.4mL
Adhesive Film,s 2 slices

Specifications

Specifications

6.3 to 100ng/mL
Human
2.2ng/mL
Quantification
Antibody coated microwells (96 wells), Biotin-Conjugate antibody (100μL), Streptavidin-HRP (150μL), Standard (lyophilized): 2 vials, Sample diluent (12mL), Assay Buffer Concentrate 20x (5mL), Wash Buffer Concentrate 20x (50mL), Substrate solution (15mL), Stop solution (15mL), Blue, green and red dye (0.4mL each)
ELISA Kit
For research use only (RUO)
10μL
Store the controls at -20°C. Others at 4°C. Avoid repeated freezing and thawing.
10μL
HRP
ICAM1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human sICAM-1.
Colorimetric
96 wells
ICAM1 (Human) ELISA Kit
1 Kit
Amniotic Fluid, Bile, Cell Culture Supernatant, Plasma (Citrate, EDTA, Heparin), Serum, Spontaneous Urine
2.2ng/mL
96 Assays
Videos
Documents