missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ICAM1 (Human) ELISA Kit

Brand:  Abnova™ KA3044.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007795

  • 578.00 EUR / 1 set


Estimated Shipment
03-09-2020
Add to basket

Includes: One 96-well plate pre-coated with anti-Human ICAM-1 antibody, 96 (8 x 12) wells
Lyophilized ICAM-1 standards, 10ng x 2
Sample/Standard diluent buffer, 30mL
Biotin conjugated anti-Human ICAM-1 antibody (Concentrated), Dilution: 1:100, 130μL
Antibody diluent buffer, 12mL
Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
ABC diluent buffer, 12mL
TMB substrate, 10mL
Stop solution, 10mL
Wash buffer (25X), 30mL

Description

Description

This kit was based on sandwich enzyme-linked immune-sorbent assay technology
Specifications

Specifications

156 to 10000pg/mL
Human
10pg/mL
Quantification
One 96-well plate pre-coated with anti-Human ICAM-1 antibody (96 (8 x 12) wells), Lyophilized ICAM-1 standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human ICAM-1 antibody (Concentrated), Dilution: 1:100: 130μL, Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X): 30mL
RUO
100μL
Store at 4°C for 4 months, or at -20°C for 8 months.
100μL
Biotin
ICAM1 (Human) ELISA Kit is intended for the quantitative measurement of human ICAM1.
Colorimetric
96 wells
ICAM1 (Human) ELISA Kit
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
<10pg/mL
96 Assays
Videos
Documents