missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Hp (Rat) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Rat Haptoglobulin in Plasma and Serum

Brand:  Abnova™ KA1403.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006625

  • 578.00 EUR / 1 set


Estimated Shipment
12-02-2020
Add to basket

Specifications

Specifications

1000 to 15000ng/mL
Rat
20ng/mL
Quantification
Hp (Rat) ELISA Kit
1 Kit
Plasma, Serum
96 Assays
Biotin
Haptoglobulin (Rat) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of rat haptoglobulin in plasma and serum.
Colorimetric
96-well Antibody coated microwell, 60μg Rat Haptoglobulin standard, 1 vial Biotinylated Haptoglobulin, 30mL MIX diluent concentrate (10X), 30mL Wash Buffer 20X, 80μL Streptavidin Peroxidase Conjugate (SP Conjugate), 8mL
HP (Rat) ELISA kit
RUO
Store the kit at 4°C or -20°C.
25μL
Videos
Documents