missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Hp (Rat) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Rat Haptoglobulin in Plasma and Serum
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Rat |
Product Type | Hp (Rat) ELISA Kit |
Quantity | 1 Kit |
Regulatory Status | For research use only (RUO) |
Specifications
Colorimetric | |
Hp (Rat) ELISA Kit | |
For research use only (RUO) |
Rat | |
1 Kit |