missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HP (Dog) ELISA Kit

Brand:  Abnova™ KA2127.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007055

  • 578.00 EUR / 1 set


Estimated Shipment
20-02-2020
Add to basket

Includes: Diluent Concentrate (Running buffer)
Wash Solution Concentrate
Enzyme-Antibody Conjugate 100X: 150μL of affinity purified anti-Dog Heptoglobin antibody conjugated with horseradish peroxidase in a stabilizing buffer
Chromogen-Substrate Solution
Stop Solution: 0.3M sulfuric acid. WARNING: Avoid contact with skin
Anti-Dog Heptoglobin ELISA Micro Plate: Twelve removable eight (8) well micro well strips in well holder frame. Each well is coated with affinity-purified anti-Dog Hp
Dog Heptoglobin Calibrator: a lyophilized Dog Heptoglobin Calibrator

Specifications

Specifications

1.95 to 125ng/mL
Dog
Colorimetric
96 wells
HP (Dog) ELISA Kit
1 Kit
Biological Fluid
Store the kit at 4°C.
100μL
HRP
HP (Dog) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of dog HP.
Quantification
Diluent concentrate (50mL), Wash solution concentrate (50mL), Enzyme-Antibody conjugate concentrate (150μL), TMB substrate solution (12mL), Stop solution (12mL), Microtiter plate (96-well), Calibrator (Lyophilized): 1 vial
ELISA Kit
RUO
100μL
96 Assays
Videos
Documents