missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HP (Dog) ELISA Kit
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | HRP (Horseradish Peroxidase) |
Cross Reactivity | Dog |
Product Type | ELISA Kit |
Regulatory Status | For research use only (RUO) |
Description
The dog haptoglobin ELISA is based on a solid phase enzymelinked immunosorbent assay (ELISA)Specifications
Colorimetric | |
Dog | |
For research use only (RUO) |
HRP (Horseradish Peroxidase) | |
ELISA Kit | |
100 μL |