missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HP (Bovine) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Bovine HP

Brand:  Abnova™ KA1850.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006885

  • 578.00 EUR / 1 set


Estimated Shipment
31-08-2020
Add to basket

Specifications

Specifications

Biotin
HP (Bovine) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of bovine HP.
Colorimetric
96-well Antibody coated microwell, 480ng Bovine Haptoglobulin standard, 1 vial Biotinylated Haptoglobulin Antibody (100X), 30mL EIA Diluent concentrate (10X), 30mL x 2, Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromogen Substrate, 12mL stop solution
HP (Bovine) ELISA kit
For research use only (RUO)
Store components of the kit at 4°C or -20°C as described in the protocol.
50μL
Bovine
3.7ng/mL
Quantification
HP (Bovine) ELISA Kit
1 Kit
Cell Culture Supernatant, Urine
96 Assays
Videos
Documents