missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HP (Bovine) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Bovine HP
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Bovine |
Product Type | HP (Bovine) ELISA kit |
For Use With (Application) | Quantification |
Quantity | 1 Kit |
Specifications
Colorimetric | |
HP (Bovine) ELISA kit | |
1 Kit |
Bovine | |
Quantification |