missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HIV-1/2 Ab ELISA Kit

Brand:  Abnova™ KA0292.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006015

  • 263.00 EUR / 1 set


Estimated Shipment
11-02-2020
Add to basket

Includes: HIV 1+2 Antigens Plate, 1 plate
Conc. HIV Antigen Conjugate, 1 bottle 0.3mL
Anti-HIV1 Positive Control, 1 bottle 1.5mL
Anti-HIV2 Positive Control, 1 bottle 1.5mL
Conjugate Diluent D, 1 bottle 1.5mL
TMB Substrate Solution A, 1 bottle 20mL
TMB Substrate Solution B, 1 bottle 12mL
2 N Sulfuric Acid, 1 bottle 12mL
Washing Solution D (20X) Concentrate, 1 bottle 110mL
Accessories: Adhesive slips, X1; Black cover, X1

Description

Description

The kit adopts the “direct sandwich principle” as the basis for the assay to detect antibodies to HIV-1 and/or HIV-2
Specifications

Specifications

HRP
Colorimetric
96 wells
HIV-1/2 Ab ELISA Kit
1 Kit
Plasma, Serum
Store the kit at 4°C.
100μL
HIV-1/2 antibody ELISA Kit is a sandwich enzyme immunoassay for the measurement of antibodies to human Immunodeficiency Virus Type 1 and/or Type 2.
Qualitative
HIV 1+2 Antigens Plate (1 plate), Conc. HIV Antigen Conjugate (0.2mL), Anti-HIV1 Positive Control (1.0mL), Anti-HIV2 Positive Control (1.0mL), HIV Negative Control (1.0mL), Conjugate Diluent D (20mL), TMB Substrate Solution A (10mL), TMB Substrate Solution B (10mL), 2N Sulfuric Acid (10mL), Washing Solution D (20X) Concentrate (100mL)
ELISA Kit
RUO
100μL
96 Assays
Videos
Documents