missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HIV-1/2 Ab ELISA Kit
POA
Specifications
Detection Method | Colorimetric |
---|---|
Conjugate | HRP |
Product Type | ELISA Kit |
Regulatory Status | For research use only (RUO) |
Sample Type | Plasma, Serum |
Description
HIV-1/2 antibody ELISA Kit is a sandwich enzyme immunoassay for the detection of HIV-1/2 antibody.Specifications
Colorimetric | |
ELISA Kit | |
Plasma, Serum |
HRP | |
For research use only (RUO) | |
Store the kit at 4°C |