Learn More
Abnova™ HBEGF (Human) ELISA Kit
In vitro enzyme-linked immunosorbent assay for the quantitative measurement of human HBEGF
POA
Specifications
Detection Method Colorimetric
Product Type HBEGF (Human) ELISA Kit
View More Specs
Description
Description
  • Sample Type: Cell Culture Supernatant, Plasma, Serum, Urine
  • Sample Volume: 100μL
Quantification
Specifications
Specifications
Colorimetric
HBEGF (Human) ELISA Kit