Learn More
Abnova™ HBEGF (Human) ELISA Kit
In vitro enzyme-linked immunosorbent assay for the quantitative measurement of human HBEGF
Brand:  Abnova™ KA3343.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16007935
  • 578.00 EUR / 1 set
Estimated Shipment
07-08-2020
Add to basket
Description
Description
  • Sample Type: Cell Culture Supernatant, Plasma, Serum, Urine
  • Sample Volume: 100μL
Quantification
Specifications
Specifications
15.6 to 1000pg/mL
HBEGF (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human HBEGF.
Quantification
HBEGF (Human) ELISA kit
Cell Culture Supernate, Plasma (EDTA, Heparin), Serum
Store the kit at-20°C.
Human
Colorimetric
HBEGF (Human) ELISA Kit
1 Kit
100μL