missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fn1 (Mouse) ELISA Kit

Sandwich enzyme immunoassay for the quantitative measurement of mouse Fn1

Brand:  Abnova™ KA0491.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006135

  • 578.00 EUR / 1 set


Estimated Shipment
12-02-2020
Add to basket

Description

Description

  • Sample Type: Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
  • Sample Volume: 100μL

Quantification

Specifications

Specifications

3.13 to 200ng/mL
Fn1 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Fn1.
Quantification
Fn1 (Mouse) ELISA kit
Cell Culture Supernatant, Plasma, Serum, Urine
Store components of the kit at 4°C or -20°C as described in the protocol.
Mouse
Colorimetric
Fn1 (Mouse) ELISA Kit
1 Kit
50μL
Videos
Documents