missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FN1 (Human) ELISA Kit

Brand:  Abnova™ KA0188.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16005925

  • 578.00 EUR / 1 set


Estimated Shipment
11-02-2020
Add to basket

Includes: Microwell Plate coated with Polyclonal Antibody (rabbit) to human Fibronectin, 96 (8 x 12) wells
Biotin-Conjugate anti-Fibronectin polyclonal antibody, 70µL
Streptavidin-HRP, 200µL
Fibronectin Standard, lyophilized, 40ng/mL upon reconstitution, 2 vials
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), 5mL x 3
Wash Buffer Concentrate 20x (PBS with 1% Tween 20), 50mL
Substrate Solution (tetramethyl-benzidine), 15mL
Stop Solution (1M Phosphoric acid), 15mL
Adhesive Films, 4 slices

Description

Description

The FN1 (Human) ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of human Fibronectin
Specifications

Specifications

0.31 to 20ng/mL
Human
0.1ng/mL
Quantification
Antibody coated microwells (96 wells), Biotin-Conjugate antibody (100μL), Streptavidin-HRP (150μL), Standard (lyophilized): 2 vials, Sample diluent (12mL), Assay Buffer Concentrate 20x (5mL), Wash Buffer Concentrate 20x (50mL), Substrate solution (15mL), Stop solution (15mL), Blue, green and red dye (0.4mL each)
ELISA Kit
For research use only (RUO)
50μL
Store the kit at 4°C.
50μL
Biotin
Fibronectin (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human fibronectin.
Colorimetric
96 wells
FN1 (Human) ELISA Kit
1 Kit
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
0.1ng/mL
96 Assays
Videos
Documents