Learn More
Abnova™ FABP3 (Dog) ELISA Kit
Brand:  Abnova™ KA2429.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16007275
  • 578.00 EUR / 1 set
Estimated Shipment
12-02-2020
Add to basket
Description
Description
FABP3 (Dog) ELISA Kit is an enzyme immunoassay for the quantitative determination of FABP3 in dog serum or plasma.
Quantification
Specifications
Specifications
FABP3 (Dog) ELISA Kit is for the quantitative measurement of FABP3 in plasma and serum.
Quantification
FABP3 (Dog) ELISA Kit
Store the kit at 4°C.