missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ F7 (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human F7

Brand:  Abnova™ KA0471.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006115

  • 578.00 EUR / 1 set


Estimated Shipment
18-02-2020
Add to basket

Specifications

Specifications

6.25 to 200ng/mL
Human
6ng/mL
Quantification
F7 (Human) ELISA Kit
1 Kit
Cell Culture Supernatant, Plasma, Serum
96 Assays
Biotin
F7 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human F7.
Colorimetric
96 Antibody coated microwell, 270ng Human FVII standard, Biotinylated FVII Antibody(100X) vial, 30mL MIX diluent concentrate (10X), 2 30mL Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate, 8mL Chromogen Substrate, 12mL stop solution
F7 (Human) ELISA kit
For research use only (RUO)
Store components of the kit at 4°C or -20°C as described in the protocol.
50μL
Videos
Documents