missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ERBB2 (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human ERBB2

Brand:  Abnova™ KA3034.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007785

  • 578.00 EUR / 1 set


Estimated Shipment
11-02-2020
Add to basket

Specifications

Specifications

62.5 to 4000pg/mL
Human
10pg/mL
Quantification
ERBB2 (Human) ELISA Kit
RUO
Store at 4°C for 4 months, or at -20°C for 8 months.
100μL
Biotin
ERBB2 (Human) ELISA Kit is intended for the quantitative measurement of human ERBB2.
Colorimetric
96-well plate pre-coated with anti-Human ErbB-2 antibody, Lyophilized ErbB-2 standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human ErbB-2 antibody (Concentrated Dilution: 1:100) (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated Dilution: 1:100) (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
96 Assays
Body Fluid, Cell Culture Supernate, Plasma, Serum, Tissue Lysate
96 Assays
Videos
Documents